missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-1 beta / IL1B Protein
A DNA sequence encoding the Homo sapiens (Human) IL-1 beta / IL1B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6214-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000567.1 | |
IL-1 beta / IL1B Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3553 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
IL1B | |
Recombinant Protein | |
E. coli | |
APVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS |