missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFNA14 / Interferon alpha-14 Protein
A DNA sequence encoding the Homo sapiens (Human) IFNA14 / Interferon alpha-14, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6200-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002163.2 | |
IFNA14 / Interferon alpha-14 Protein | |
Research Use Only | |
IFNA14 | |
Recombinant Protein | |
Yeast | |
CNLSQTHSLN NRRTLMLMAQ MRRISPFSCL KDRHDFEFPQ EEFDGNQFQK AQAISVLHEM MQQTFNLFST KNSSAAWDET LLEKFYIELF QQMNDLEACV IQEVGVEETP LMNEDSILAV KKYFQRITLY LMEKKYSPCA WEVVRAEIMR SLSFSTNLQK RLRRKD | |
Greater than 80% as determined by SDS-PAGE. |
3448 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |