missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSPH1 Protein
A cDNA sequence encoding the HSPH1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12338-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HSPH1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMSVV GLDVGSQSCY IAVARAGGIE TIANEFSDRC TPSVISFGSK NRTIGVAAKN QQITHANNTV SNFKRFHGRA FNDPFIQKEK ENLSYDLVPL KNGGVGIKVM YMGEEHLFSV EQITAMLLTK LKETAENSLK KPVTDCVISV PSFFTDAERR SVLDAAQIVG LNCLRLMNDM TAVALNYGIY KQDLPSLDEK PRIVVFVDMG HSAFQVSACA FNKGKLKVLG TAFDPFLGGK NFDEKLVEHF CAEFKTKYKL DAKSKIRALL RLYQECEKLK KLMSSNSTDL PLNIECFMND KDVSGKMNRS QFEELCAELL QKIEVPLYSL LEQTHLKVED VSAVEIVGGA TRIPAVKERI AKFFGKDIST TLNADEAVAR GCALQCAILS PAFKVREFSV TDAVPFPISL IWNHDSEDTE GVHEVFSRNH AAPFSKVLTF LRRGPFELEA FYSDPQGVPY PEAKIGRFVV QNVSAQKDGE KSRVKVKVRV NTHGIFTIST ASMVEKVPTE ENEMSSEADM ECLNQRPPEN PDTDKNVQQD NSEAGTQPQV QTDAQQTSQS PPSPELTSEE NKIPDADKAN EKKVDQPPEA KKPKIKVVNV ELPIEANLVW QLGKDLLNMY IETEGKMIMQ DKLEKERNDA KNAVEEYVYE FRDKLCGPYE KFICEQDHQN FLRLLTETED WLYEEGEDQA KQAYVDKLEE LMKIGTPVKV RFQEAEERPK MFEELGQRLQ HYAKIAADFR NKDEKYNHID ESEMKKVEKS VNEVMEWMNN VMNAQAKKSL DQDPVVRAQE IKTKIKELNN TCEPVVTQPK PKIESPKLER TPNGPNIDKK EEDLEDKNNF GAEPPHQNGE CYPNEKNSVN MDLD | |
Greater than 90.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HSP105 protein solution contains 20mM Tris-HCl, pH 8 and 50mM NaCl. |