missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGB1 / HMG1 Protein
A DNA sequence encoding the Homo sapiens (Human) HMGB1 / HMG1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8377-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001300821.1 | |
HMGB1 / HMG1 Protein | |
Research Use Only | |
HMGB1 | |
Recombinant Protein | |
E. coli | |
KPRGKMSSYA FFVQTCREEH KKKHPDASVN FSEFSKKCSE RWKTMSAKEK GKFEDMAKAD KARYEREMKT YIPPKGETKK KFKDPNAPKR PPSAFFLFCS EYRPKIKGEH PGLSIGDVAK KLGEMWNNTA ADDKQPYEKK AAKLKEKYEK DIAAYRAKGK PDAAKKGVVK AE | |
Greater than 90% as determined by SDS-PAGE. |
3146 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |