missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-G Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-G, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6167-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002118.1 | |
HLA-G Protein | |
Research Use Only | |
HLA-G | |
Recombinant Protein | |
E. coli | |
GSHSMRYFSA AVSRPGRGEP RFIAMGYVDD TQFVRFDSDS ACPRMEPRAP WVEQEGPEYW EEETRNTKAH AQTDRMNLQT LRGYYNQSEA SSHTLQWMIG CDLGSDGRLL RGYEQYAYDG KDYLALNEDL RSWTAADTAA QISKRKCEAA NVAEQRRAYL EGTCVEWLHR YLENGKEMLQ RADPPKTHVT HHPVFDYEAT LRCWALGFYP AEIILTWQRD GEDQTQDVEL VETRPAGDGT FQKWAAVVVP SGEEQRYTCH VQHEGLPEPL MLRWKQSSLP TIPIMGIVAG LVVLAAVVTG AAVAAVLWRK KSS | |
Greater than 90% as determined by SDS-PAGE. |
3135 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |