missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-DQA2 Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-DQA2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8816-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_064440.1 | |
HLA-DQA2 Protein | |
Research Use Only | |
HLA-DQA2 | |
Recombinant Protein | |
E. coli | |
EDIVADHVAS YGVNFYQSHG PSGQYTHEFD GDEEFYVDLE TKETVWQLPM FSKFISFDPQ SALRNMAVGK HTLEFMMRQS NSTAATNEVP EVTVFSKFPV TLGQPNTLIC LVDNIFPPVV NITWLSNGHS VTEGVSETSF LSKSDHSFFK ISYLTFLPSA DEIYDCKVEH WGLDEPLLKH WEPEIPAPMS E | |
Greater than 90% as determined by SDS-PAGE. |
3118 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |