missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone H3.3 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone H3.3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6141-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002098.1 | |
Histone H3.3 Protein | |
Research Use Only | |
H3F3A | |
Recombinant Protein | |
E. coli | |
ARTKQTARKS TGGKAPRKQL ATKAARKSAP STGGVKKPHR YRPGTVALRE IRRYQKSTEL LIRKLPFQRL VREIAQDFKT DLRFQSAAIG ALQEASEAYL VGLFEDTNLC AIHAKRVTIM PKDIQLARRI RGERA | |
Greater than 90% as determined by SDS-PAGE. |
3020 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |