Learn More
enQuireBio™ Recombinant Human Hexokinase-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Hexokinase-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6165-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000179.2 | |
Hexokinase-1 Protein | |
Research Use Only | |
HK1 | |
Recombinant Protein | |
E. coli | |
HFHLTKDMLL EVKKRMRAEM ELGLRKQTHN NAVVKMLPSF VRRTPDGTEN GDFLALDLGG TNFRVLLVKI RSGKKRTVEM HNKIYAIPIE IMQGTGEELF DHIVSCISDF LDYMGIKGPR MPLGFTFSFP CQQTSLDAGI LITWTKGFKA TDCVGHDVVT LLRDAIKRRE EFDLDVVAVV NDTVGTMMTC AYEEPTCEVG LIVGTGSNAC YMEEMKNVEM VEGDQGQMCI NMEWGAFGDN GCLDDIRTHY DRLVDEYSLN AGKQRYEKMI SGMYLGEIVR NILIDFTKKG FLFRGQISET LKTRGIFETK FLSQIESDRL ALLQVRAILQ QLGLNSTCDD SILVKTVCGV VSRRAAQLCG AGMAAVVDKI RENRGLDRLN VTVGVDGTLY KLHPHFSRIM HQTVKELSPK CNVSFLLSED GSGKGAALIT AVGVRLRTEA SS | |
Greater than 90% as determined by SDS-PAGE. |
3098 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |