Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein Q, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8440-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001153145.1 | |
Heterogeneous nuclear ribonucleoprotein 1 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10492 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
NSAP1 | |
Recombinant Protein | |
E. coli | |
ATEHVNGNGT EEPMDTTSAV IHSENFQTLL DAGLPQKVAE KLDEIYVAGL VAHSDLDERA IEALKEFNED GALAVLQQFK DSDLSHVQNK SAFLCGVMKT YRQREKQGTK VADSSKGPDE AKIKALLERT GYTLDVTTGQ RKYGGPPPDS VYSGQQPSVG TEIFVGKIPR DLFEDELVPL FEKAGPIWDL |