missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HBEGF / DTR Protein
A DNA sequence encoding the Human HBEGF / DTR Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10400-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001936.1 | |
HBEGF / DTR Protein | |
Research Use Only | |
HBEGF | |
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 1 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 130 mM NaCl, lyophilized |
1839 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
DLQEADLDLL RVTLSSKPQA LATPNKEEHG KRKKKGKGLG KKRDPCLRKY KDFCIHGECK YVKELRAPSC ICHPGYHGER CHGLSL | |
Greater than 97% as determined by SDS-PAGE and HPLC. |