missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HADHB, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) HADHB, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8435-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000174.1 | |
HADHB, mitochondrial Protein | |
Research Use Only | |
HADHB | |
Recombinant Protein | |
E. coli | |
APAVQTKTKK TLAKPNIRNV VVVDGVRTPF LLSGTSYKDL MPHDLARAAL TGLLHRTSVP KEVVDYIIFG TVIQEVKTSN VAREAALGAG FSDKTPAHTV TMACISANQA MTTGVGLIAS GQCDVIVAGG VELMSDVPIR HSRKMRKLML DLNKAKSMGQ RLSLISKFRF NFLAPELPAV SEFSTSETMG HSADRLAAAF AVSRLEQDEY ALRSHSLAKK AQDEGLLSDV VPFKVPGKDT VTKDNGIRP | |
Greater than 90% as determined by SDS-PAGE. |
3032 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |