missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Growth Hormone Receptor / GHR / GHBP Protein
A DNA sequence encoding the Homo sapiens (Human) Growth Hormone Receptor / GHR / GHBP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6083-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000154.1 | |
Growth Hormone Receptor / GHR / GHBP Protein | |
Research Use Only | |
GHR | |
Recombinant Protein | |
E. coli | |
FSGSEATAAI LSRAPWSLQS VNPGLKTNSS KEPKFTKCRS PERETFSCHW TDEVHHGTKN LGPIQLFYTR RNTQEWTQEW KECPDYVSAG ENSCYFNSSF TSIWIPYCIK LTSNGGTVDE KCFSVDEIVQ PDPPIALNWT LLNVSLTGIH ADIQVRWEAP RNADIQKGWM VLEYELQYKE VNETKWKMMD PILTTSVPVY SLKVDKEYEV RVRSKQRNSG NYGEFSEVLY VTLPQMSQFT CEEDFY | |
Greater than 81.9% as determined by SDS-PAGE. |
2690 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |