missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Growth / differentiation factor 9 Protein
A DNA sequence encoding the Homo sapiens (Human) Growth/differentiation factor 9, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6076-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001275753.1 | |
Growth / differentiation factor 9 Protein | |
Research Use Only | |
GDF9 | |
Recombinant Protein | |
E. coli | |
GQETVSSELK KPLGPASFNL SEYFRQFLLP QNECELHDFR LSFSQLKWDN WIVAPHRYNP RYCKGDCPRA VGHRYGSPVH TMVQNIIYEK LDSSVPRPSC VPAKYSPLSV LTIEPDGSIA YKEYEDMIAT KCTCR | |
Greater than 90% as determined by SDS-PAGE. |
2661 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |