Learn More
enQuireBio™ Recombinant Human GPD1 / Glycerolphosphate Dehydrogenase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) GPD1 / Glycerolphosphate Dehydrogenase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6111-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001244128.1 | |
GPD1 / Glycerolphosphate Dehydrogenase 1 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2819 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
GPD1 | |
Recombinant Protein | |
E. coli | |
MASKKVCIVG SGNWGSAIAK IVGGNAAQLA QFDPRVTMWV FEEDIGGKKL TEIINTQHEN VKYLPGHKLP PNVVAVPDVV QAAEDADILI FVVPHQFIGK ICDQLKGHLK ANATGISLIK GVDEGPNGLK LISEVIGERL GIPMSVLMGA NIASEVADEK FCETTIGCKD PAQGQLLKEL MQTPNFRITV VQEVDTVEIC GALKNVVAVG AGFCDGLGFG DNTKAAVIRL GLMEMIAFAK LFCSGPVSSA TFLESCGVAD LITTCYGGRN RKVAEAFART GKSIEQLEKE LLNGQKLQGP ETARELYSIL QHKGLVDKFP LFMAVYKVCY EGQPVGEFIH CLQNHPEHM |