missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GP2 Protein
A cDNA sequence encoding the GP2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12031-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GP2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MPHLMERMVG SGLLWLALVS CILTQASAVQ RGYGNPIEAS SYGLDLDCGA PGTPEAHVCF DPCQNYTLLD EPFRSTENSA GSQGCDKNMS GWYRFVGEGG VRMSETCVQV HRCQTDAPMW LNGTHPALGD GITNHTACAH WSGNCCFWKT EVLVKACPGG YHVYRLEGTP WCNLRYCTVP RDPSTVEDKC EKACRPEEEC LALNSTWGCF CRQDLNSSDV HSLQPQLDCG PREIKVKVDK CLLGGLGLGE EVIAYLRDPN CSSILQTEER NWVSVTSPVQ ASACRNILER NQTHAIYKNT LSLVNDFIIR DTILNINFQC AYPLDMKVSL QAALQPIVSS LNVSVDGNGE FIVRMALFQD QNYTNPYEGD AVELSVESVL YVGAILEQGD TSRFNLVLRN CYATPTEDKA DLVKYFIIRN SCSNQRDSTI HVEENGQSSE SRFSVQMFMF AGHYDLVFLH CEIHLCDSLN EQCQPSCSRS QVRSEVPAID LARVLDLGPI TRRGAQSPGV MNGTPSTAGF LVAWPMVLLT VLLAWLF | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GP2 solution is supplied in 16mM HEPES buffer pH 7.5, 80mM NaCl and 20% glycerol. |