missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glycophorin A / CD235a Protein
A DNA sequence encoding the Homo sapiens (Human) Glycophorin A / CD235a, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6135-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001295119.1 | |
Glycophorin A / CD235a Protein | |
Research Use Only | |
GYPA | |
Recombinant Protein | |
Yeast | |
SSTTGVAMHT STSSSVTKSY ISSQTNDTHK RDTYAATPRA HEVSEISVRT VYPPEEETGE RVQLAHHFSE PE | |
Greater than 90% as determined by SDS-PAGE. |
2993 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |