missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GGCT Protein
A DNA sequence encoding the Homo sapiens (Human) GGCT, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6080-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001186744.1 | |
GGCT Protein | |
Research Use Only | |
GGCT | |
Recombinant Protein | |
E. coli | |
MANSGCKDVT GPDEESFLYF AYGSNLLTER IHLRNPSAAF FCVARLQDFK LDFGNSQGKT SQTWHGGIAT IFQSPGDEVW GVVWKMNKSN LNSLDEQEGV KSGMYVVIEV KVATQEGKEI TCRSYLMTNY ESAPPSPQYK KIICMGAKEN GLPLEYQEKL KAIEPNDYTG KVSEEIEDII KKGETQTL | |
Greater than 90% as determined by SDS-PAGE. |
79017 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |