missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDNF Protein
A DNA sequence encoding the Homo sapiens (Human) GDNF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6078-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000505.1 | |
GDNF Protein | |
Research Use Only | |
GDNF | |
Recombinant Protein | |
E. coli | |
SPDKQMAVLP RRERNRQAAA ANPENSRGKG RRGQRGKNRG CVLTAIHLNV TDLGLGYETK EELIFRYCSG SCDAAETTYD KILKNLSRNR RLVSDKVGQA CCRPIAFDDD LSFLDDNLVY HILRKHSAKR CGCI | |
Greater than 90% as determined by SDS-PAGE. |
2668 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |