missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GADD45G / CR6 Protein
A DNA sequence encoding the Homo sapiens (Human) GADD45G / CR6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6064-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006696.1 | |
GADD45G / CR6 Protein | |
Research Use Only | |
GADD45G | |
Recombinant Protein | |
E. coli | |
MTLEEVRGQD TVPESTARMQ GAGKALHELL LSAQRQGCLT AGVYESAKVL NVDPDNVTFC VLAAGEEDEG DIALQIHFTL IQAFCCENDI DIVRVGDVQR LAAIVGAGEE AGAPGDLHCI LISNPNEDAW KDPALEKLSL FCEESRSVND WVPSITLPE | |
Greater than 90% as determined by SDS-PAGE. |
10912 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |