missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GABRA4 Protein
A DNA sequence encoding the Homo sapiens (Human) GABRA4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6061-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000800.2 | |
GABRA4 Protein | |
Research Use Only | |
GABRA4 | |
Recombinant Protein | |
E. coli | |
QNQKEEKLCT ENFTRILDSL LDGYDNRLRP GFGGPVTEVK TDIYVTSFGP VSDVEMEYTM DVFFRQTWID KRLKYDGPIE ILRLNNMMVT KVWTPDTFFR NGKKSVSHNM TAPNKLFRIM RNGTILYTMR LTISAECPMR LVDFPMDGHA CPLKFGSYAY PKSEMIYTWT KGPEKSVEVP KESSSLVQYD LIGQTVSSET IKSITGEYIV MTVYFHLRRK MGY | |
Greater than 90% as determined by SDS-PAGE. |
2557 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |