missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human G antigen 5 Protein
A DNA sequence encoding the Homo sapiens (Human) G antigen 5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6065-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001465.1 | |
G antigen 5 Protein | |
Research Use Only | |
GAGE5 | |
Recombinant Protein | |
E. coli | |
MSWRGRSTYY WPRPRRYVQP PEVIGPMRPE QFSDEVEPAT PEEGEPATQR QDPAAAQEGE DEGASAGQGP KPEADSQEQG HPQTGCECED GPDGQEMDPP NPEEVKTPEE GEKQSQC | |
Greater than 90% as determined by SDS-PAGE. |
2576 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |