missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FZD4 Protein
A cDNA sequence encoding the FZD4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11914-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FZD4 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPFGDEEER RCDPIRISMC QNLGYNVTKM PNLVGHELQT DAELQLTTFT PLIQYGCSSQ LQFFLCSVYV PMCTEKINIP IGPCGGMCLS VKRRCEPVLK EFGFAWPESL NCSKFPPQND HNHMCMEGPG DEEVPLPHKT PIQPGEECHS VGTNSDQYIW VKRSLNCVLK CGYDAGLYSRSAKEFTDIWL EPKSCDKTHT CPPCPAPELL GGPSVFLFPP KPKDTLMISR TPEVTCVVVD VSHEDPEVKF NWYVDGVEVH NAKTKPREEQ YNSTYRVVSV LTVLHQDWLN GKEYKCKVSN KALPAPIEKT ISKAKGQPRE PQVYTLPPSR DELTKNQVSL TCLVKGFYPS DIAVEWESNGQPENNYKTTP PVLDSDGSFF LYSKLTVDKS RWQQGNVFSC SVMHEALHNH YTQKSLSLSP GKHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FZD4 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |