missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FUT3 Protein
A cDNA sequence encoding the FUT3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11911-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FUT3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRVSRDDA TGSPRAPSGS SRQDTTPTRP TLLILLWTWP FHIPVALSRC SEMVPGTADC HITADRKVYP QADTVIVHHW DIMSNPKSRL PPSPRPQGQR WIWFNLEPPP NCQHLEALDR YFNLTMSYRS DSDIFTPYGW LEPWSGQPAH PPLNLSAKTE LVAWAVSNWK PDSARVRYYQ SLQAHLKVDV YGRSHKPLPK GTMMETLSRY KFYLAFENSL HPDYITEKLW RNALEAWAVP VVLGPSRSNY ERFLPPDAFI HVDDFQSPKD LARYLQELDK DHARYLSYFR WRETLRPRSF SWALDFCKAC WKLQQESRYQ TVRSIAAWFT | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FUT3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |