missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fractalkine / CX3CL1 Protein
A DNA sequence encoding the Homo sapiens (Human) Fractalkine / CX3CL1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5896-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001291321.1 | |
Fractalkine / CX3CL1 Protein | |
Research Use Only | |
CX3CL1 | |
Recombinant Protein | |
E. coli | |
QHHGVTKCNI TCSKMTSKIP VALLIHYQQN QASCGKRAII LETRQHRLFC ADPKEQWVKD AMQHLDRQAA ALTRNG | |
Greater than 90% as determined by SDS-PAGE. |
6376 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |