missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FLT4 Fc Protein

A cDNA sequence encoding the FLT4 Fc was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10619-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15994428

  • 7146.73€ / 1 miligramo
Envío estimado: 01-03-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Especificaciones

Especificaciones

FLT4 Fc Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Insect
MQRGAALCLR LWLCLGLLDG LVSGYSMTPP TLNITEESHV IDTGDSLSIS CRGQHPLEWA WPGAQEAPAT GDKDSEDTGV VRDCEGTDAR PYCKVLLLHE VHANDTGSYV CYYKYIKARI EGTTAASSYV FVRDFEQPFI NKPDTLLVNR KDAMWVPCLV SIPGLNVTLR SQSSVLWPDG QEVVWDDRRG MLVSTPLLHD ALYLQCETTW GDQDFLSNPF LVHITGNELY DIQLLPRKSL ELLVGEKLVL NCTVWAEFNS GVTFDWDYPG KQAERGKWVP ERRSQQTHTE LSSILTIHNV SQHDLGSYVC KANNGIQRFR ESTEVIVHEN PFISVEWLKG PILEATAGDE LVKLPVKLAA YPPPEFQWYK DGKALSGRHS PHALVLKEVT EASTGTYTLA LWNSAAGLRR NISLELVVNV PPQIHEKEAS SPSIYSRHSR QALTCTAYGV PLPLSIQWHW RPWTPCKMFA QRSLRRRQQQ DLMPQCRDWR AVTTQDAVNP IESLDTWTEF VEGKNKTVSK LVIQNANVSA MYKCVVSNKV GQDERLIYFY VTTIPDGFTI ESKPSEELLE GQPVLLSCQA DSYKYEHLRW YRLNLSTLHD AHGNPLLLDC KNVHLFATPL AASLEEVAPG ARHATLSLSI PRVAPEHEGH YVCEVQDRRS HDKHCHKKYL SVQALEAPRL TQNLTDLLVN VSDSLEMQCL VAGAHAPSIV WYKDERLLEE KSGVDLADSN QKLSIQRVRE EDAGRYLCSV CNAKGCVNSS ASVAVEGSED KGSMEIVILV GTGVIAVFFW VLLLLIFCNM RRPAHADIKT GYLSIIMDPG EVPLEEQCEY LSYDASQWEF PRERLHLGRV LGYGAFGKVV EASAFGIHKG SSCDTVAVKM LKEGATASEH RALMSELKIL IHIGNHLNVV NLLGACTKPQ GPLMVIVEFC KYGNLSNFLR AKRDAFSPCA EKSPEQRGRF RAMVELARLD RRRPGSSDRV LFARFSKTEG GARRASPDQE AEDLWLSPLT MEDLVCYSFQ VARGMEFLAS RKCIHRDLAA RNILLSESDV VKICDFGLAR DIYKDPDYVR KGSARLPLKW MAPESIFDKV YTTQSDVWSF GVLLWEIFSL GASPYPGVQI NEEFCQRLRD GTRMRAPELA TPAIRRIMLN CWSGDPKARP AFSELVEILG DLLQGRGLQE EEEVCMAPRS SQSSEEGSFS QVSTMALHIA QADAEDSPPS LQRHSLAARY YNWVSFPGCL ARGAETRGSS RMKTFEEFPM TPTTYKGSVD NQTDSGMVLA SEEFEQIESR HRQESGFSCK GPGQNVAVTR AHPDSQGRRR RPERGARGGQ VFYNSEYGEL SEPSEEDHCS PSARVTFFTD NSY
Greater than 90.0% as determined by(a)Analysis by RP-HPLC. (b)Analysis by SDS-PAGE.
Research Use Only
Measured by its ability to bind recombinant rat VEGF-C in a functional solid phase binding assay. Immobilised recombinant human VEGFR-3/FLT-4 at 5 microg/ml can bind recombinant rat VEGF-C in a linear range of 8-500 ng/ml.
Human
Untagged
FLT4 Fc Chimera was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives.
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales