missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FKBP14 Protein
A DNA sequence encoding the Homo sapiens (Human) FKBP14, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8076-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_060416.1 | |
FKBP14 Protein | |
Research Use Only | |
FKBP14 | |
Recombinant Protein | |
E. coli | |
ALIPEPEVKI EVLQKPFICH RKTKGGDLML VHYEGYLEKD GSLFHSTHKH NNGQPIWFTL GILEALKGWD QGLKGMCVGE KRKLIIPPAL GYGKEGKGKI PPESTLIFNI DLLEIRNGPR SHESFQEMDL NDDWKLSKDE VKAYLKKEFE KHGAVVNESH HDALVEDIFD KEDEDKDGFI SAREFTYKHD EL | |
Greater than 90% as determined by SDS-PAGE. |
55033 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |