missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fibroleukin Protein
A DNA sequence encoding the Homo sapiens (Human) Fibroleukin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7571-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006673.1 | |
Fibroleukin Protein | |
Research Use Only | |
FGL2 | |
Recombinant Protein | |
E. coli | |
NNETEEIKDE RAKDVCPVRL ESRGKCEEAG ECPYQVSLPP LTIQLPKQFS RIEEVFKEVQ NLKEIVNSLK KSCQDCKLQA DDNGDPGRNG LLLPSTGAPG EVGDNRVREL ESEVNKLSSE LKNAKEEINV LHGRLEKLNL VNMNNIENYV DSKVANLTFV VNSLDGKCSK CPSQEQIQSR PVQHLIYKDC SDYYAIGKRS SETYRVTPDP KNSSFEVYCD METMGGGWTV LQARLDGSTN FTRTWQDYKA GFGNLRREFW LGNDKIHLLT KSKEMILRID LEDFNGVELY ALYDQFYVAN EFLKYRLHVG NYNGTAGDAL RFNKHYNHDL KFFTTPDKDN DRYPSGNCGL YYSSGWWFDA CLSANLNGKY YHQKYRGVRN GIFWGTWPGV SEAHPGGYKS SFKEAKMMIR PKHFKP | |
Greater than 90% as determined by SDS-PAGE. |
10875 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |