missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fibroblast growth factor 13 Protein
A DNA sequence encoding the Homo sapiens (Human) Fibroblast growth factor 13, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6042-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001132970.1 | |
Fibroblast growth factor 13 Protein | |
Research Use Only | |
FGF13 | |
Recombinant Protein | |
E. coli | |
MAAAIASSLI RQKRQARERE KSNACKCVSS PSKGKTSCDK NKLNVFSRVK LFGSKKRRRR RPEPQLKGIV TKLYSRQGYH LQLQADGTID GTKDEDSTYT LFNLIPVGLR VVAIQGVQTK LYLAMNSEGY LYTSELFTPE CKFKESVFEN YYVTYSSMIY RQQQSGRGWY LGLNKEGEIM KGNHVKKNKP AAHFLPKPLK VAMYKEPSLH DLTEFSRSGS GTPTKSRSVS GVLNGGKSMS HNEST | |
Greater than 90% as determined by SDS-PAGE. |
2258 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |