missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGFR4 Protein
A cDNA sequence encoding the FGFR4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10606-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
2264 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Determined by its ability to inhibit human FGF acidic-dependent proliferation on R1 cells. The ED50 for this effect is typically at 15.0-30.0 ng/ml. | |
Human | |
Untagged | |
CD334 was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives. |
FGFR4 Protein | |
Research Use Only | |
FGFR4 | |
Recombinant Protein | |
Baculovirus | |
MRLLLALLGV LLSVPGPPVL SLEASEEVEL EPCLAPSLEQ QEQELTVALG QPVRLCCGRA ERGGHWYKEG SRLAPAGRVR GWRGRLEIAS FLPEDAGRYL CLARGSMIVL QNLTLITGDS LTSSNDDEDP KSHRDPSNRH SYPQQAPYWT HPQRMEKKLH AVPAGNTVKF RCPAAGNPTP TIRWLKDGQA FHGENRIGGI RLRHQHWSLV MESVVPSDRG TYTCLVENAV GSIRYNYLLD VLERSPHRPI LQAGLPANTT AVVGSDVELL CKVYSDAQPH IQWLKHIVIN GSSFGADGFP YVQVLKTADI NSSEVEVLYL RNVSAEDAGE YTCLAGNSIG LSYQSAWLTV LPEEDPTWTA AAPEARYTDI ILYASGSLAL AVLLLLAGLY RGQALHGRHP RPPATVQKLS RFPLARQFSL ESGSSGKSSS SLVRGVRLSS SGPALLAGLV SLDLPLDPLW EFPRDRLVLG KPLGEGCFGQ VVRAEAFGMD PARPDQASTV AVKMLKDNAS DKDLADLVSE MEVMKLIGRH KNIINLLGVC TQEGPLYVIV ECAAKGNLRE FLRARRPPGP DLSPDGPRSS EGPLSFPVLV SCAYQVARGM QYLESRKCIH RDLAARNVLV TEDNVMKIAD FGLARGVHHI DYYKKTSNGR LPVKWMAPEA LFDRVYTHQS DVWSFGILLW EIFTLGGSPY PGIPVEELFS LLREGHRMDR PPHCPPELYG LMRECWHAAP SQRPTFKQLV EALDKVLLAV SEEYLDLRLT FGPYSPSGGD ASSTCSSSDS VFSHDPLPLG SSSFPFGSGV QT | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |