missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF20 Protein
A cDNA sequence encoding the FGF20 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10603-3ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FGF20 Protein | |
Research Use Only | |
The ED50, as measured in a proliferation assay using mouse NR6R-3T3 cells, is less than 2.5ng/ml. | |
Human | |
His | |
Lyophilized from a 0.2microm filtered solution in MOPS, (NH4)2SO4, DTT and EDTA. |
3 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MHHHHHHAPL AEVGGFLGGL EGLGQQVGSH FLLPPAGERP PLLGERRSAA ERSARGGPGA AQLAHLHGIL RRRQLYCRTG FHLQILPDGS VQGTRQDHSL FGILEFISVA VGLVSIRGVD SGLYLGMNDK GELYGSEKLT SECIFREQFE ENWYNTYSSN IYKHGDTGRR YFVALNKDGT PRDGARSKRH QKFTHFLPRP VDPERVPELY KDLLMYT | |
Greater than 97.0% as determined by SDS-PAGE. |