missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF12 Protein
A DNA sequence encoding the Homo sapiens (Human) FGF12, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6041-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004104.3 | |
FGF12 Protein | |
Research Use Only | |
FGF12 | |
Recombinant Protein | |
E. coli | |
MESKEPQLKG IVTRLFSQQG YFLQMHPDGT IDGTKDENSD YTLFNLIPVG LRVVAIQGVK ASLYVAMNGE GYLYSSDVFT PECKFKESVF ENYYVIYSST LYRQQESGRA WFLGLNKEGQ IMKGNRVKKT KPSSHFVPKP IEVCMYREPS LHEIGEKQGR SRKSSGTPTM NGGKVVNQDS T | |
Greater than 80% as determined by SDS-PAGE. |
2257 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |