missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF10 / KGF2 Protein
A DNA sequence encoding the Homo sapiens (Human) FGF10 / KGF2 Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6038-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004456.1 | |
FGF10 / KGF2 Protein | |
Research Use Only | |
FGF10 | |
Recombinant Protein | |
E. coli | |
QALGQDMVSP EATNSSSSSF SSPSSAGRHV RSYNHLQGDV RWRKLFSFTK YFLKIEKNGK VSGTKKENCP YSILEITSVE IGVVAVKAIN SNYYLAMNKK GKLYGSKEFN NDCKLKERIE ENGYNTYASF NWQHNGRQMY VALNGKGAPR RGQKTRRKNT SAHFLPMVVH S | |
Greater than 80% as determined by SDS-PAGE. |
2255 | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |