missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF 18 Protein
A cDNA sequence encoding the FGF 18 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11869-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FGF 18 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASEENVDFRIHV ENQTRARDDV SRKQLRLYQL YSRTSGKHIQ VLGRRISARG EDGDKYAQLL VETDTFGSQV RIKGKETEFY LCMNRKGKLV GKPDGTSKEC VFIEKVLENN YTALMSAKYS GWYVGFTKKG RPRKGPKTRE NQQDVHFMKR YPKGQPELQK PFKYTTVTKR SRRIRPTHPA | |
Purity as determined by densitometric image analysis is greater than 95%. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FGF18 was filtered (0.4microm) and lyophilized in phosphate buffered saline and 5% w/v trehalose. |