missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FDX1 Protein
A cDNA sequence encoding the FDX1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11864-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FDX1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MASMTGGQQM GRGSMSSSED KITVHFINRD GETLTTKGKV GDSLLDVVVE NNLDIDGFGA CEGTLACSTC HLIFEDHIYE KLDAITDEEN DMLDLAYGLT DRSRLGCQIC LTKSMDNMTV RVPETVADAR QSIDVGKTS | |
Greater than 90.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The FDX1 solution contains 20mM Tris-HCl buffer (pH 8.0) and 10% Glycerol. |