missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FcERI / FCER1A Protein
A DNA sequence encoding the Homo sapiens (Human) FcERI / FCER1A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6026-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001992.1 | |
FcERI / FCER1A Protein | |
Research Use Only | |
FCER1A | |
Recombinant Protein | |
Yeast | |
VPQKPKVSLN PPWNRIFKGE NVTLTCNGNN FFEVSSTKWF HNGSLSEETN SSLNIVNAKF EDSGEYKCQH QQVNESEPVY LEVFSDWLLL QASAEVVMEG QPLFLRCHGW RNWDVYKVIY YKDGEALKYW YENHNISITN ATVEDSGTYY CTGKVWQLDY ESEPLNITVI KAPREKYWLQ | |
Greater than 90% as determined by SDS-PAGE. |
2205 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |