missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fas Ligand / FASLG / CD95L Protein
A DNA sequence encoding the Homo sapiens (Human) Fas Ligand / FASLG / CD95L, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6021-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000630.1 | |
Fas Ligand / FASLG / CD95L Protein | |
Research Use Only | |
FASLG | |
Recombinant Protein | |
E. coli | |
QIGHPSPPPE KKELRKVAHL TGKSNSRSMP LEWEDTYGIV LLSGVKYKKG GLVINETGLY FVYSKVYFRG QSCNNLPLSH KVYMRNSKYP QDLVMMEGKM MSYCTTGQMW ARSSYLGAVF NLTSADHLYV NVSELSLVNF EESQTFFGLY KL | |
Greater than 90% as determined by SDS-PAGE. |
356 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |