missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP7 Protein
A cDNA sequence encoding the FABP7 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11837-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FABP7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MVEAFCATWK LTNSQNFDEY MKALGVGFAT RQVGNVTKPT VIISQEGDKV VIRTLSTFKN TEISFQLGEE FDETTADDRN CKSVVSLDGD KLVHIQKWDG KETNFVREIK DGKMVMTLTF GDVVAVRHYE KA | |
Greater than 95.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The FABP7 protein solution contains 25mM Tris-HCl pH 7.5, 2mM EDTA and 10% Glycerol. |