missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP6 / I-BABP Protein
A DNA sequence encoding the Homo sapiens (Human) FABP6 / I-BABP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6015-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Specifications
NP_001035532.1 | |
FABP6 / I-BABP Protein | |
Research Use Only | |
FABP6 | |
Recombinant Protein | |
E. coli | |
MAFTGKFEME SEKNYDEFMK LLGISSDVIE KAHNFKIVTE VQQDGQDFTW SQHYYGGHTM TNKFTVGKES NIQTMGGKTF KATVQMEGGK LVVNFPNYHQ TSEIVGDKLV EVSTIGGVTY ERVSKRLA | |
Greater than 80% as determined by SDS-PAGE. |
2172 | |
200 μg | |
Not determined. | |
Full Length of BC022489 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |