missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP2 Protein
A cDNA sequence encoding the FABP2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11834-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FABP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
AFDSTWKVDR SENYDKFMEK MGVNIVKRKL AAHDNLKLTI TQEGNKFTVK ESSAFRNIEV VFELGVTFNY NLADGTELRG TWSLEGNKLI GKFKRTDNGN ELNTVREIIG DELVQTYVYE GVEAKRIFKK D | |
Greater than 97.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
FABP2 was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |