missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F-actin-capping protein subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) F-actin-capping protein subunit beta, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5760-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001193469.1 | |
F-actin-capping protein subunit beta Protein | |
Research Use Only | |
CAPZB | |
Recombinant Protein | |
E. coli | |
MSDQQLDCAL DLMRRLPPQQ IEKNLSDLID LVPSLCEDLL SSVDQPLKIA RDKVVGKDYL LCDYNRDGDS YRSPWSNKYD PPLEDGAMPS ARLRKLEVEA NNAFDQYRDL YFEGGVSSVY LWDLDHGFAG VILIKKAGDG SKKIKGCWDS IHVVEVQEKS SGRTAHYKLT STVMLWLQTN KSGSGTMNLG GSLTRQMEKD ETVSDCSPHI ANIGRLVEDM ENKIRSTLNE IYFGKTKDIV NGLRSVQTFA DKSKQEALKN DLVEALKRKQ QC | |
Greater than 80% as determined by SDS-PAGE. |
832 | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |