missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Extracellular calcium-sensing receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Extracellular calcium-sensing receptor, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5764-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000379.2 | |
Extracellular calcium-sensing receptor Protein | |
Research Use Only | |
CASR | |
Recombinant Protein | |
E. coli | |
YGPDQRAQKK GDIILGGLFP IHFGVAAKDQ DLKSRPESVE CIRYNFRGFR WLQAMIFAIE EINSSPALLP NLTLGYRIFD TCNTVSKALE ATLSFVAQNK IDSLNLDEFC NCSEHIPSTI AVVGATGSGV STAVANLLGL FYIPQVSYAS SSRLLSNKNQ FKSFLRTIPN DEHQATAMAD IIEYFRWNWV GTIAADDDYG RPGIEKFREE AEERDICIDF SELISQYSDE EEIQHVVEVI QNSTAKVIVV FSSGPDLEPL IKEIVRRNIT GKIWLASEAW ASSSLIAMPQ YFHVVGGTIG FALKAGQIPG FREFLKKVHP RKSVHNGFAK EFWEETFNCH LQEGAKGPLP VDTFLRGHEE SGDRFSNSST AFRPLCTGDE NISSVETPYI DYTHLRISYN VYLAVYSIAH ALQDIYTCLP GRGLFTNGSC ADIKKVEAWQ VLKHLRHLNF TNNMGEQVTF DECGDLVGNY SIINWHLSPE DGSIVFKEVG YYNVYAKKGE RLFINEEKIL WSGFSREVPF SNCSRDCLAG TRKGIIEGEP TCCFECVECP DGEYSDETDA SACNKCPDDF WSNENHTSCI AKEIEFLSWT EPF | |
Greater than 90% as determined by SDS-PAGE. |
846 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |