missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Exonuclease V Protein
A DNA sequence encoding the Homo sapiens (Human) Exonuclease V, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8066-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001333875.1 | |
Exonuclease V Protein | |
Research Use Only | |
EXO5 | |
Recombinant Protein | |
E. coli | |
MAETREEETV SAEASGFSDL SDSEFLEFLD LEDAQESKAL VNMPGPSSES LGKDDKPISL QNWKRGLDIL SPMERFHLKY LYVTDLATQN WCELQTAYGK ELPGFLAPEK AAVLDTGASI HLARELELHD LVTVPVTTKE DAWAIKFLNI LLLIPTLQSE GHIREFPVFG EGEGVLLVGV IDELHYTAKG ELELAELKTR RRPMLPLEAQ KKKDCFQVSL YKYIFDAMVQ GKVTPASLIH HTKLCLEKPL GPSVLRHAQQ GGFSVKSLGD LMELVFLSLT LSDLPVIDIL KIEYIHQETA TVLGTEIVAF KEKEVRAKVQ HYMAYWMGHR EPQGVDVEEA WKCRTCTYAD ICEWRKGSGV LSSTLAPQVK KAK | |
Greater than 90% as determined by SDS-PAGE. |
64789 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |