missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ETS1 Protein
A cDNA sequence encoding the ETS1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11814-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
2113 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKAAVDLKPT LTIIKTEKVD LELFPSPDME CADVPLLTPS SKEMMSQALK ATFSGFTKEQ QRLGIPKDPR QWTETHVRDW VMWAVNEFSL KGVDFQKFCM NGAALCALGK DCFLELAPDF VGDILWEHLE ILQKEDVKPY QVNGVNPAYP ESRYTSDYFI SYGIEHAQCV PPSEFSEPSF ITESYQTLHP ISSEELLSLK YENDYPSVIL RDPLQTDTLQ NDYFAIKQEV VTPDNMCMGR TSRGKLGGQD SFESIESYDS CDRLTQSWSS QSSFNSLQRV PSYDSFDSED YPAALPNHKP KGTFKDYVRD RADLNKDKPV IPAAALAGYT GSGPIQLWQF LLELLTDKSC QSFISWTGDG WEFKLSDPDE VARRWGKRKN KPKMNYEKLS RGLRYYYDKN IIHKTAGKRY VYRFVCDLQS LLGYTPEELH AMLDVKPDAD E |
ETS1 Protein | |
Research Use Only | |
ETS1 | |
Human | |
Untagged | |
ETS1 protein in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |