missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ES1 protein homolog, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ES1 protein homolog, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8386-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004640.4 | |
ES1 protein homolog, mitochondrial Protein | |
Research Use Only | |
C21orf33 | |
Recombinant Protein | |
E. coli | |
ARVALVLSGC GVYDGTEIHE ASAILVHLSR GGAEVQIFAP DVPQMHVIDH TKGQPSEGES RNVLTESARI ARGKITDLAN LSAANHDAAI FPGGFGAAKN LSTFAVDGKD CKVNKEVERV LKEFHQAGKP IGLCCIAPVL AAKVLRGVEV TVGHEQEEGG KWPYAGTAEA IKALGAKHCV KEVVEAHVDQ KNKVVTTPAF MCETALHYIH DGIGAMVRKV LELTGK | |
Greater than 90% as determined by SDS-PAGE. |
102724023 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |