missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Erythropoietin / EPO Protein
A DNA sequence encoding the Human Erythropoietin / EPO, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10349-ma-37500IU
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000790.2 | |
Erythropoietin / EPO Protein | |
Research Use Only | |
EPO | |
The Specific Activity was measured by the stimulation of reticulocyte production in normocyth-aemic mice and was found to be no less than 1.5 M 105 IU/mg. | |
Human | |
Untagged | |
0.2um filtered sodium citrate buffer (1 liter of ddH2O containing 5.9 g of sodium citrate, 5.8 g of sodium chloride and 0.06 g of citric acid), lyophilized |
2056 | |
37500 IU | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
Mammal | |
APPRLICDSR VLERYLLEAK EAENITTGCA EHCSLNENIT VPDTKVNFYA WKRMEVGQQA VEVWQGLALL SEAVLRGQAL LVNSSQPWEP LQLHVDKAVS GLRSLTTLLR ALGAQKEAIS PPDAASAAPL RTITADTFRK LFRVYSNFLR GKLKLYTGEA CRTGDR | |
Greater than 98% as determined by SDS-PAGE and HPLC. |