missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ENHO Protein
A cDNA sequence encoding the ENHO was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11787-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
ENHO Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
ENHO filtered (0.4 microm) and lyophilized from 0.5 mg/ml in phosphate buffered saline. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGGKSNGEKK YIVGFKQGFK SCAKKEDVIS EKGGKLQKCF KYVDAASATL NEKAVEELKK DPSVAYVEED KLFKALTSCHSRSADVDSLS ESSPNSSPGP CPEKAPPPQK PSHEGSYLLQ P |