missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Elongation factor Tu, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Elongation factor Tu, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6855-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_003312.3 | |
Elongation factor Tu, mitochondrial Protein | |
Research Use Only | |
TUFM | |
Recombinant Protein | |
E. coli | |
AVEAKKTYVR DKPHVNVGTI GHVDHGKTTL TAAITKILAE GGGAKFKKYE EIDNAPEERA RGITINAAHV EYSTAARHYA HTDCPGHADY VKNMITGTAP LDGCILVVAA NDGPMPQTRE HLLLARQIGV EHVVVYVNKA DAVQDSEMVE LVELEIRELL TEFGYKGEET PVIVGSALCA LEGRDPELGL KSVQKLLDAV DTYIPVPARD LEKPFLLPVE AVYSVPGRGT VVTGTLERGI LKKGDECELL GHSKNIRTVV TGIEMFHKSL ERAEAGDNLG ALVRGLKRED LRRGLVMVKP GSIKPHQKVE AQVYILSKEE GGRHKPFVSH FMPVMFSLTW DMACRIILPP EKELAMPGED LKFNLILRQP MILEKGQRFT LRDGNRTIGT GLVTNTLAMT EEEKNIKWG | |
Greater than 90% as determined by SDS-PAGE. |
7284 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |