missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF4H Protein
A cDNA sequence encoding the EIF4H was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11774-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EIF4H Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMADFDT YDDRAYSSFG GGRGSRGSAG GHGSRSQKEL PTEPPYTAYV GNLPFNTVQG DIDAIFKDLS IRSVRLVRDK DTDKFKGFCY VEFDEVDSLK EALTYDGALL GDRSLRVDIA EGRKQDKGGF GFRKGGPDDR GMGSSRESRG GWDSRDDFNS GFRDDFLGGR GGSRPGDRRT GPPMGSRFRD GPPLRGSNMD FREPTEEERA QRPRLQLKPR TVATPLNQVA NPNSAIFGGA RPREEVVQKE QE | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
EIF4H protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |