missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF4A3 Protein
A cDNA sequence encoding the EIF4A3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11770-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EIF4A3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMATTAT MATSGSARKR LLKEEDMTKV EFETSEEVDV TPTFDTMGLR EDLLRGIYAY GFEKPSAIQQ RAIKQIIKGR DVIAQSQSGT GKTATFSISV LQCLDIQVRE TQALILAPTR ELAVQIQKGL LALGDYMNVQ CHACIGGTNV GEDIRKLDYG QHVVAGTPGR VFDMIRRRSL RTRAIKMLVL DEADEMLNKG FKEQIYDVYR YLPPATQVVL ISATLPHEIL EMTNKFMTDP IRILVKRDEL TLEGIKQFFV AVEREEWKFD TLCDLYDTLT ITQAVIFCNT KRKVDWLTEK MREANFTVSS MHGDMPQKER ESIMKEFRSG ASRVLISTDV WARGLDVPQV SLIINYDLPN NRELYIHRIG RSGRYGRKGV AINFVKNDDI RILRDIEQYY STQIDEMPMN VADLI | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
EIF4A3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 30% glycerol and 200mM NaCl. |