missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF3E Protein
A DNA sequence encoding the Homo sapiens (Human) EIF3E, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5971-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001559.1 | |
EIF3E Protein | |
Research Use Only | |
EIF3E | |
Recombinant Protein | |
E. coli | |
AEYDLTTRIA HFLDRHLVFP LLEFLSVKEI YNEKELLQGK LDLLSDTNMV DFAMDVYKNL YSDDIPHALR EKRTTVVAQL KQLQAETEPI VKMFEDPETT RQMQSTRDGR MLFDYLADKH GFRQEYLDTL YRYAKFQYEC GNYSGAAEYL YFFRVLVPAT DRNALSSLWG KLASEILMQN WDAAMEDLTR LKETIDNNSV SSPLQSLQQR TWLIHWSLFV FFNHPKGRDN IIDLFLYQPQ YLNAIQTMCP HILRYLTTAV ITNKDVRKRR QVLKDLVKVI QQESYTYKDP ITEFVECLYV NFDFDGAQKK LRECESVLVN DFFLVACLED FIENARLFIF ETFCRIHQCI SINMLADKLN MTPEEAERWI VNLIRNARLD AKIDSKLGHV VMGNNAVSPY QQVIEKTKSL SFRSQMLAMN IEKKLNQNSR SEAPNWATQD SGFY | |
Greater than 80% as determined by SDS-PAGE. |
3646 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |